Learn More
Cytiva Membranes
Brand: Cytiva 10409870
Code : A0
Additional Details : Weight : 0.01000kg
Specifications
0.6 μm | |
These membranes are used to quantitatively evaluate microorganism counts. The liquid is filtered through a membrane and microorganisms collect on the membrane surface. | |
Membrane |
Sterile | |
100Each |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Warning
Hazard Category
- Flammable solid Category 1
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose