Learn More
Carl Zeiss™ MembraneSlide
Product Code. 15511306
Click to view available options
:
MembraneSlide 1.0 PET (D)
Product Code. 15511306
Brand: Carl Zeiss™ 4151909051000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Ready to use PET membrane-covered slide with blue marking area. Zeiss MembraneSlides provide excellent DNA, RNA or protein recovery. Available in packs of 100.
MembraneSlides are glass slides covered with a thin membrane which is biochemically inert. Membrane will be cut with the sample and acts as a stabilizing backbone during the lifting process. Therefore, a single laser pulse can lift even a large area without affecting the morphological integrity. This feature is especially important for the isolation of single cells, chromosomes as well as for live cells or small organisms. Also visualization of collected samples in the cap is facilitated.
Compatible Microscopes:
  • PALM CombiSystem Rel. 4.2
  • PALM MicroBeam Rel.4.2
  • PALM MicroTweezers Rel.4.2
Specifications
Material PET Membrane-covered Glass
Diameter (Metric) 1.0 mm
Quantity 100 Slides