missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Menidazole, Honeywell Fluka™
Click to view available options
Quantity:
10 mg
Description
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
- High quality inorganic and organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
- Free expert technical support
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Physical Form | Solid |
CAS | 696-23-1 |
Grade | Analytical Standard |
Packaging | Glass Bottle |
Density | 1.43 g/cm3 |
Quantity | 10 mg |
Flash Point | 195.4°C |
For Use With (Application) | Antibiotics |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'