Learn More
Invitrogen™ Mer (MERTK) Human ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EHMERX5
Description
The Human Mer (MERTK) ELISA quantitates Hu Mer in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu Mer. Principle of the method The HumanM er solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
MERTK is a receptor tyrosine kinase that transduces signals from the extracellular matrix into the cytoplasm by binding to several ligands including LGALS3, TUB, TULP1 or GAS6. Regulates many physiological processes including cell survival, migration, differentiation, and phagocytosis of apoptotic cells (efferocytosis). Ligand binding at the cell surface induces autophosphorylation of MERTK on its intracellular domain that provides docking sites for downstream signaling molecules. Following activation by ligand, interacts with GRB2 or PLCG2 and induces phosphorylation of MAPK1, MAPK2, FAK/PTK2 or RAC1. MERTK signaling plays a role in various processes such as macrophage clearance of apoptotic cells, platelet aggregation, cytoskeleton reorganization and engulfment. Functions in the retinal pigment epithelium (RPE) as a regulator of rod outer segments fragments phagocytosis. Plays also an important role in inhibition of Toll-like receptors (TLRs)-mediated innate immune response by activating STAT1, which selectively induces production of suppressors of cytokine signaling SOCS1 and SOCS3.Specifications
Q12866 | |
0.15 ng/mL | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
5 x 96 assays | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
10461 | |
<12% | |
Standard, Assay Diluent Concentrate, Biotinylated Detection Antibody, Chromogen, Adhesive Plate Covers, Wash Buffer, Stop Solution, SAV-HRP, Pre-coated 96 well plates | |
5 x 96 Tests | |
Apoptosis | |
2°C to 8°C | |
Human | |
4 hr. 45 min. |
0.164 ng/mL to 40 ng/mL | |
0.15 ng/mL to 40 ng/mL | |
Biotin | |
Kinase & Inhibitors | |
This ELISA kit shows no cross-reactivity with the following cytokines tested: ANGPTL3, beta IG-H3, CA9, Cathepsin B, CD23, CHI3L1, CTLA-4 (CD152), DKK-4, DPPIV (CD26), EDA-A2, Erythropoietin R, FGF-6, FGF-9, Gas 1, GITR (TNFRSF18), IGFBP-5, IL-1 F5 (FIL1 delta), IL-1 F6 (FIL1 epsilon), IL-1 F7 (FIL1 zeta), IL-1 F8 (FIL1 eta), IL-1 F9 (IL-1 H1), IL-1 F10 (IL-1 HY2), IL-1 R5, IL-21 (IL-17 C), IL-18, IL-20, IL-34, IL-5 R alpha, IL-10 R alpha, Layilin, Leptin R, Marapsin, MMP-7, P-Cadherin, Prostasin, PSMA (FOLH1), SIGIRR, TGF beta R3, Coagulation Factor III (Tissue Factor, F3), TWEAK (TNFSF12) | |
Human | |
Colorimetric Microplate Reader | |
MERTK | |
<10% | |
HRP | |
RUO | |
Plasma, 50 μ;L; Serum, 50 μ;L; Supernatant, 100 μ;L | |
MER, RP38, Tyro12, c-Eyk, c-mer | |
1 hr. 20 min. |