missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mercaptoacetic acid sodium salt, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Glass bottle
Plastic bottle
Description
Reagent that protects tryptophan in amino acid analysis
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | >300°C |
Packaging | Plastic bottle |
Linear Formula | HSCH2CO2Na |
Quantity | 25 g |
UN Number | UN2811 |
Beilstein | 4569109 |
Sensitivity | Air sensitive; Hygroscopic |
Formula Weight | 114.1 |
Percent Purity | 98% |
Physical Form | Powder |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'