missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mercaptoacetic acid, sodium salt, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | >300.0°C |
Assay Percent Range | 97.5% min. (Iodimetry) |
Packaging | Glass bottle |
Linear Formula | HSCH2CO2Na |
Quantity | 250 g |
Merck Index | 15,8819 |
Solubility Information | Solubility in water: 1000g/L (20°C). Other solubilities: slightly soluble in alcohol |
Formula Weight | 114.09 |
Percent Purity | 98% |
Chemical Name or Material | Mercaptoacetic acid, sodium salt, 98% |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0460742 | Certificate of Analysis | 01/03/2024 | 148892500,148890500,148890010 |
A0458901 | Certificate of Analysis | 22/12/2023 | 148892500,148890500,148890010 |
A0453504 | Certificate of Analysis | 25/08/2023 | 148892500,148890500,148890010 |
A0437795 | Certificate of Analysis | 25/08/2023 | 148892500,148890500,148890010 |
A0395660 | Certificate of Analysis | 15/12/2022 | 148892500,148890500,148890010 |
Safety and Handling


Product Identifier
- Mercaptoacetic acid, sodium salt
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- Substances and mixtures corrosive to metals Category 1
- Respiratory sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H301-Toxic if swallowed.
- H312-Harmful in contact with skin.
- H290-May be corrosive to metals.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P390-Absorb spillage to prevent material damage.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Mercaptoacetic acid, sodium salt, 98%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction