missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mercuric Chloride solution EU Pharmacopoeia, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
10 mL
Packaging:
Ampoule
Unit Size:
1000mL
Specifications
Specifications
| Quantity | 10 mL |
| Purity Grade Notes | EU Ph Ref: 1052201 |
| Grade | Eu. Ph. |
| Concentration or Composition (by Analyte or Components) | 10μg/mL |
| Packaging | Ampoule |
Safety and Handling
Product Identifier
- Mercuric Chloride Solution. European Pharmacopoeia Ref: 1052201 (RP 119)
- Danger
- Acute toxicity Category 2
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 2
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H300-Fatal if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H341-Suspected of causing genetic defects.
- H361-Suspected of damaging fertility or the unborn child.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
- P202-Do not handle until all safety precautions have been read and understood.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P361 + P364-Take off immediately all contaminated clothing and wash it before reuse.
- P501-Dispose of contents/container to .
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction