missing translation for 'onlineSavingsMsg'
Learn More

Mercurochrome, 24-27% Mercury, Thermo Scientific Chemicals

Product Code. 10308670
100 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129-16-8
Molecular Formula C20H8Br2HgNa2O6
Molecular Weight (g/mol) 750.65
MDL Number MFCD00013081
InChI Key QJJYLIJNUFJILQ-UHFFFAOYSA-N
Synonym Merbromin, 2, 7-Dibromo-4-hydroxymercurifluoresceine disodium salt
PubChem CID 54598522
IUPAC Name sodium;[2,7-dibromo-9-(2-carboxyphenyl)-3-hydroxy-6-oxoxanthen-4-yl]mercury;hydrate
SMILES C1=CC=C(C(=C1)C2=C3C=C(C(=O)C=C3OC4=C(C(=C(C=C24)Br)O)[Hg])Br)C(=O)O.O.[Na+]

Specifications

Melting Point >300°C
Color Green
Packaging Glass bottle
Quantity 100 g
Merck Index 15, 5934
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in alcohol,acetone,ether
Formula Weight 750.65
Percent Purity 24 to 27% (Hg), 18 to 22.5% (Br2)
Physical Form Glimmering Scales, Granules or Crystals
Chemical Name or Material Mercurochrome
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Mercurochrome, 24-27% Mercury
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H373-May cause damage to organs.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P361-Take off immediately all contaminated clothing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Mercurochrome, 24-27% Mercury, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.