missing translation for 'onlineSavingsMsg'
Learn More
Mercury, Honeywell™ Product Code.: 15647270

Mercury, Honeywell™

Product Code. 15647270
100 g
Click to view available options
Quantity:
100 g
This item is not returnable. View return policy

Product Code. 15647270

Brand: Honeywell Research Chemicals 83359100G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 7439-97-6
Molecular Formula Hg
MDL Number MFCD00011035
UN Number UN2809
Quantity 100 g
Synonym hydrargyrum, quicksilver, metallic, liquid silver, mercurio, quecksilber, mercure, quick silver, colloidal, mercury, metallic
InChI Key QSHDDOUJBYECFT-UHFFFAOYSA-N
SMILES [Hg]
IUPAC Name mercury
Molecular Weight (g/mol) 200.59
PubChem CID 23931
ChEBI CHEBI:16170
Formula Weight 200.59g/mol
Percent Purity 99%
Vapor Pressure 0.01 mmHg (20°C), 1 mmHg (126°C)
Chemical Name or Material Mercury
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Mercury, Honeywell™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.