missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) Acetate, Certified AR for Analysis, Fisher Chemical™

£118.72 - £511.00

Chemical Identifiers

CAS 1600-27-7
Molecular Formula C4H6HgO4
Molecular Weight (g/mol) 318.68
MDL Number 12165
InChI Key BRMYZIKAHFEUFJ-UHFFFAOYSA-L
IUPAC Name mercury(2+) diacetate
SMILES [Hg++].CC([O-])=O.CC([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10101770
View Documents
Fisher Chemical
M/2652/47
50 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10735721
View Documents
Fisher Chemical
M/2652/50
250 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

1600-27-7
318.68
BRMYZIKAHFEUFJ-UHFFFAOYSA-L
[Hg++].CC([O-])=O.CC([O-])=O
C4H6HgO4
12165
mercury(2+) diacetate

Specifications

179°C
White
Powder/Solid
12165
[Hg++].CC([O-])=O.CC([O-])=O
318.68
Plastic powder jar
1600-27-7
4.4
C4H6HgO4
BRMYZIKAHFEUFJ-UHFFFAOYSA-L
mercury(2+) diacetate
318.67g/mol
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Mercuric acetate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H310-Fatal in contact with skin.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H300-Fatal if swallowed.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P361-Take off immediately all contaminated clothing.

SDS
Documents

Documents

Product Certifications