missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) Acetate, Extra Pure, SLR, Fisher Chemical

Product Code. 10508250
250g, Plastic powder jar
Click to view available options
250g, Plastic powder jar
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Melting Point 179°C
CAS 1600-27-7
Color White
pH 4.4
Physical Form Powder/Solid
Quantity 250 g
Molecular Formula C4H6HgO4
MDL Number 12165
InChI Key BRMYZIKAHFEUFJ-UHFFFAOYSA-L
SMILES [Hg++].CC([O-])=O.CC([O-])=O
IUPAC Name mercury(2+) diacetate
Molecular Weight (g/mol) 318.68
Formula Weight 318.67g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Mercuric acetate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H310-Fatal in contact with skin.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P361-Take off immediately all contaminated clothing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Mercury(II) Acetate, Extra Pure, SLR, Fisher Chemical >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.