missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) bromide, 99+%

Product Code. 10501261 Shop All Thermo Scientific Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Product Code. 10501261

Brand: Thermo Scientific Acros 190491000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

CAS: 7789-47-1 | Br2Hg | 360.4 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 7789-47-1
Molecular Formula Br2Hg
Molecular Weight (g/mol) 360.4
MDL Number MFCD00011040
InChI Key NGYIMTKLQULBOO-UHFFFAOYSA-L
Synonym mercuric bromide, mercury dibromide, mercury ii bromide, hgbr2, mercury bromide hgbr2, mercuric dibromide, mercury ii bromide 1:2, hsdb 829, hg2, mercury 2 bromide
PubChem CID 24612
ChEBI CHEBI:49639
IUPAC Name dibromomercury
SMILES Br[Hg]Br

Spécification

Boiling Point 322.0°C
Melting Point 236.0°C
Color White
Physical Form Fine Crystalline Powder
Quantity 100 g
Assay Percent Range 99% min
Linear Formula HgBr2
Fieser 05,324
Merck Index 15, 5942
Solubility Information Solubility in water: 0.6g/100mL (25 c). Other solubilities: freely soluble in hcl and hbr, soluble in alcohol and ether, slightly soluble in chloroform
Formula Weight 360.4
Percent Purity ≥99%
Packaging Glass bottle
Density 6.1
Chemical Name or Material Mercury(II) bromide, p.a.
Afficher plus Afficher moins
compliance-icons compliance-icons compliance-icons
Identifiant produit
  • Mercury(II) bromide
Mention d'avertissement
  • Danger
Catégorie de danger
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Mention de danger
  • H310-Fatal in contact with skin.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H300-Fatal if swallowed.
Conseils de prudence
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis