missing translation for 'onlineSavingsMsg'
Learn More
Mercury(II) chloride, 99.5%, Thermo Scientific Chemicals Product Code.: 10134170

Mercury(II) chloride, 99.5%, Thermo Scientific Chemicals

Product Code. 10134170
100 g, Plastic bottle
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Product Code. 10134170

Brand: Thermo Scientific Acros 201431000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7487-94-7
Molecular Formula Cl2Hg
Molecular Weight (g/mol) 271.49
MDL Number MFCD00011041
InChI Key LWJROJCJINYWOX-UHFFFAOYSA-L
Synonym mercuric chloride, mercury ii chloride, sublimate, mercury bichloride, mercury dichloride, corrosive sublimate, mercuric bichloride, mercury perchloride, sublimat, hgcl2Show MoreShow Less
PubChem CID 24085
ChEBI CHEBI:31823
IUPAC Name dichloromercury
SMILES [Cl-].[Cl-].[Hg++]

Specifications

Boiling Point 302.0°C
Melting Point 277.0°C
Color White
Physical Form Crystalline Powder and/or Crystals
Quantity 100 g
Assay Percent Range 99.4% min. (Complexometry)
Linear Formula HgCl2
Fieser 01,652; 05,427; 06,359; 09,291; 11,467; 12,304; 13,175; 15,200
Merck Index 15, 5943
Solubility Information Solubility in water: 7.4 g/100 mL (20°C). Other solubilities: 1g/40 mL acetic acid, 1g/200 mL benzene - 1g/3.8 mL alcohol, 1g/22 mL ether - 1g/12 mL glycerol, soluble in acetone and ethyl acetate, slightly soluble in pyridine and cs2
Formula Weight 271.5
Percent Purity 99.5%
Packaging Plastic bottle
Density 5.44
Chemical Name or Material Mercury(II) chloride, p.a.
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463318 Certificate TypeCertificate of Analysis Date14/06/2024 Catalog Number201430250,201435000,201431000,201430010
Lot NumberA0445009 Certificate TypeCertificate of Analysis Date09/02/2024 Catalog Number201430250,201435000,201431000,201430010
Lot NumberA0415579 Certificate TypeCertificate of Analysis Date17/01/2023 Catalog Number201430250,201435000,201431000,201430010
Lot NumberA0401038 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number201430250,201435000,201431000,201430010
Lot NumberA0398267 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number201430250,201435000,201431000,201430010
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Mercury(II) chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H300-Fatal if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H341-Suspected of causing genetic defects.
  • H361f-Suspected of damaging fertility.
  • H372-Causes damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 25908

    Expires2025-03-31

    Days

    4

    Hours

    15

    Minutes

    28

    Seconds

    32

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Mercury(II) chloride, 99.5%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.