missing translation for 'onlineSavingsMsg'
Learn More
Mercury(II) chloride, ACS, 99.5% min, Thermo Scientific Chemicals Product Code.: 11321619

Mercury(II) chloride, ACS, 99.5% min, Thermo Scientific Chemicals

Product Code. 11321619
500 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11321619

Brand: Thermo Scientific Alfa Aesar 012274.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mercury(II) chloride is used as a catalyst in making plastics, depolarizer in batteries and as a reagent in organic synthesis and analytical chemistry. It is also used to prepare an amalgam with metals such as aluminum, which finds application as reducing agent in organic synthesis. It also finds application in photography as a photographic intensifier. Further, it serves as a preservative and used in the treatment of syphilis. It is also used to remove dithiane groups bonded to a carbonyl in an umpolung reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7487-94-7
Molecular Formula Cl2Hg
Molecular Weight (g/mol) 271.49
MDL Number MFCD00011041
InChI Key LWJROJCJINYWOX-UHFFFAOYSA-L
Synonym mercuric chloride, mercury ii chloride, sublimate, mercury bichloride, mercury dichloride, corrosive sublimate, mercuric bichloride, mercury perchloride, sublimat, hgcl2
PubChem CID 24085
ChEBI CHEBI:31823
IUPAC Name dichloromercury
SMILES [Cl-].[Cl-].[Hg++]

Specifications

Boiling Point 302°C
Melting Point 280°C to 282°C
Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range 99.5% min.
Solubility Information Soluble in boiling water (solubility in cold water is increased by HCl or alkali chlorides),alcohol,glycerol,methanol,acetone,and ethyl acetateSoluble in water,alcohol,acetone and ethyl acetate. Slightly soluble in benzene,carbon disulfide and pyridine.
Formula Weight 271.5
Grade ACS
Odor Odorless
Refractive Index 1.859
Appearance White powder
Vapor Pressure 0.0001 hPa
Density 5.4
Chemical Name or Material Mercury(II) chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN28L048 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog Number012274.14,012274.36,012274.22
Lot NumberY18K022 Certificate TypeCertificate of Analysis Date20/11/2024 Catalog Number012274.14,012274.36,012274.22
Lot NumberX11K035 Certificate TypeCertificate of Analysis Date30/10/2024 Catalog Number012274.14,012274.36,012274.22
Lot NumberU06K071 Certificate TypeCertificate of Analysis Date22/08/2024 Catalog Number012274.14,012274.36,012274.22
Lot NumberR08K077 Certificate TypeCertificate of Analysis Date17/05/2024 Catalog Number012274.14,012274.36,012274.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Mercury(II) chloride, ACS, 99.5% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.