missing translation for 'onlineSavingsMsg'
Learn More

Mercury(Ii) Chloride, puriss. p.a. ACS Reagent, Honeywell Fluka™

Puriss. p.a., ACS Reagent, Reag. ISO,≥99.5% (calc. to the dried substance)

£108.36 - £452.03

Chemical Identifiers

CAS 7487-94-7
Molecular Formula Cl2Hg
Molecular Weight (g/mol) 271.49
MDL Number MFCD00011041
InChI Key LWJROJCJINYWOX-UHFFFAOYSA-L
Synonym mercuric chloride, mercury ii chloride, sublimate, mercury bichloride, mercury dichloride, corrosive sublimate, mercuric bichloride, mercury perchloride, sublimat, hgcl2
PubChem CID 24085
ChEBI CHEBI:31823
IUPAC Name dichloromercury
SMILES [Cl-].[Cl-].[Hg++]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15683150
View Documents
Honeywell-Fluka
31005-100G
100 g
£108.36
100g
Please to purchase this item. Need a web account? Register with us today!
15693150
View Documents
Honeywell-Fluka
31005-500G
500 g
£452.03
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

7487-94-7
271.49
LWJROJCJINYWOX-UHFFFAOYSA-L
24085
dichloromercury
Cl2Hg
MFCD00011041
mercuric chloride, mercury ii chloride, sublimate, mercury bichloride, mercury dichloride, corrosive sublimate, mercuric bichloride, mercury perchloride, sublimat, hgcl2
CHEBI:31823
[Cl-].[Cl-].[Hg++]

Specifications

7487-94-7
HgCl2
1624
LWJROJCJINYWOX-UHFFFAOYSA-L
dichloromercury
24085
271.5g/mol
ACS/puriss. p.a.
Mercury(Ii) Chloride, Reag. ISO
Cl2Hg
MFCD00011041
mercuric chloride, mercury ii chloride, sublimate, mercury bichloride, mercury dichloride, corrosive sublimate, mercuric bichloride, mercury perchloride, sublimat, hgcl2
[Cl-].[Cl-].[Hg++]
271.49
CHEBI:31823
≥99.5% (calc. to the dried substance)
1.3mmHg (236°C)
Videos
Safety and Handling

Safety and Handling

P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338

H300 + H310-H314-H341-H361f-H372-H410

missing translation for 'einecsNumber' : 231-299-8

missing translation for 'rtecsNumber' : OV9100000

Product Identifier
  • Mercury(II) chloride

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 1

Hazard Statement
  • H300-Fatal if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H341-Suspected of causing genetic defects.
  • H361f-Suspected of damaging fertility.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P234-Keep only in original container.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : mercury dichloride

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.