Learn More
Mercury(II) iodide, 99+%, Thermo Scientific Chemicals
£77.49 - £303.51
Chemical Identifiers
CAS | 7774-29-0 |
---|---|
Molecular Formula | HgI2 |
Molecular Weight (g/mol) | 454.401 |
MDL Number | MFCD00011044 |
InChI Key | YFDLHELOZYVNJE-UHFFFAOYSA-L |
Synonym | mercury diiodide, mercuric iodide, mercury ii iodide, red mercuric iodide, mercuric iodide, red, mercury biniodide, mercury iodide, mercurius bijodatus, hydrargyrum diodatum, hydrargyrum bijodatum |
PubChem CID | 24485 |
ChEBI | CHEBI:49659 |
IUPAC Name | diiodomercury |
SMILES | I[Hg]I |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11483167
|
Thermo Scientific Alfa Aesar
A16130.22 |
100 g |
£77.49
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11493167
|
Thermo Scientific Alfa Aesar
A16130.36 |
500 g |
£303.51
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mercury(II) iodide is is used for preparation of Nessler's reagent, used for detection of presence of ammonia. It is a semiconductor material, used in some x-ray and gamma ray detection and imaging devices operating at room temperatures. In veterinary medicine, it is used in blister ointments in exostoses, bursal enlargement, etc.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMercury(II) iodide is is used for preparation of Nessler′s reagent, used for detection of presence of ammonia. It is a semiconductor material, used in some x-ray and gamma ray detection and imaging devices operating at room temperatures. In veterinary medicine, it is used in blister ointments in exostoses, bursal enlargement, etc.
Solubility
Insoluble in water. Slightly soluble in alcohol, ether, acetone, chloroform, ethyl acetate, olive oil and castor oil.
Notes
Light sensitive. Store away from oxidizing agents.
Chemical Identifiers
7774-29-0 | |
454.401 | |
YFDLHELOZYVNJE-UHFFFAOYSA-L | |
24485 | |
diiodomercury |
HgI2 | |
MFCD00011044 | |
mercury diiodide, mercuric iodide, mercury ii iodide, red mercuric iodide, mercuric iodide, red, mercury biniodide, mercury iodide, mercurius bijodatus, hydrargyrum diodatum, hydrargyrum bijodatum | |
CHEBI:49659 | |
I[Hg]I |
Specifications
350°C (sublimation) | |
7774-29-0 | |
MFCD00011044 | |
14,5879 | |
Insoluble in water. Slightly soluble in alcohol,ether,acetone,chloroform,ethyl acetate,olive oil and castor oil. | |
I[Hg]I | |
454.401 | |
CHEBI:49659 | |
≥99% | |
Light sensitive | |
Mercury(II) iodide |
259°C | |
HgI2 | |
UN1638 | |
mercury diiodide, mercuric iodide, mercury ii iodide, red mercuric iodide, mercuric iodide, red, mercury biniodide, mercury iodide, mercurius bijodatus, hydrargyrum diodatum, hydrargyrum bijodatum | |
YFDLHELOZYVNJE-UHFFFAOYSA-L | |
diiodomercury | |
24485 | |
454.4 | |
Odorless | |
6.28 |
Safety and Handling
GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c
H300+H310+H330-H373
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MERCURY IODIDE
missing translation for 'einecsNumber' : 231-873-8
missing translation for 'rtecsNumber' : OW5250000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Mercury(II) iodide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 1
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H373-May cause damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P330-Rinse mouth.
Supplemental information
- MIXTURE LIST-Contains : Mercuric iodide
RUO – Research Use Only