missing translation for 'onlineSavingsMsg'
Learn More
Mercury(II) iodide, ACS, 99.0% min (Assay-dried basis), Thermo Scientific Chemicals Product Code.: 11381639

Mercury(II) iodide, ACS, 99.0% min (Assay-dried basis), Thermo Scientific Chemicals

Product Code. 11381639
100 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11381639

Brand: Thermo Scientific Alfa Aesar 012289.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Use As An Analytical Reagent For the Preparation of Nessler'S Reagent.

As an analytical reagent for the preparation of Nessler's Reagent. It is also used as an oxidizing agent for the synthesis of quinoxaline. Polycrystalline films of mercury (II) iodide, also play an important role in X-ray imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7774-29-0
Molecular Formula HgI2
Molecular Weight (g/mol) 454.401
MDL Number MFCD00011044
InChI Key YFDLHELOZYVNJE-UHFFFAOYSA-L
Synonym mercury diiodide, mercuric iodide, mercury ii iodide, red mercuric iodide, mercuric iodide, red, mercury biniodide, mercury iodide, mercurius bijodatus, hydrargyrum diodatum, hydrargyrum bijodatumShow MoreShow Less
PubChem CID 24485
ChEBI CHEBI:49659
IUPAC Name diiodomercury
SMILES I[Hg]I

Specifications

Boiling Point 350°C (sublimation)
Melting Point 259°C
Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.0% min.
Solubility Information Insoluble in water. Soluble in boiling alcohol,ether,acetone,ethyl acetate,carbon disulfide,alkali iodides.
Formula Weight 454.4
Grade ACS
Odor Odorless
Sensitivity Light sensitive
Appearance Red powder or beads
Density 6.28 g/cm3 at 20°C
Chemical Name or Material Mercury(II) iodide, Assay-dried basis
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberF21Y007 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number012289.22,012289.36
Lot NumberC23X040 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number012289.22,012289.36
Lot NumberZ01D061 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number012289.22,012289.36
Lot NumberR06A036 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number012289.22,012289.36
Lot NumberX13G024 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number012289.22,012289.36
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Mercury(II) iodide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
Supplemental information
  • MIXTURE LIST-Contains : Mercuric iodide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Mercury(II) iodide, ACS, 99.0% min (Assay-dried basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.