missing translation for 'onlineSavingsMsg'
Learn More
Mercury(II) iodide, ACS reagent, Red, Thermo Scientific Chemicals Product Code.: 12307213

Mercury(II) iodide, ACS reagent, Red, Thermo Scientific Chemicals

Product Code. 12307213
500 g, Glass Bottle
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 12307213

Brand: Thermo Scientific Acros 193505000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7774-29-0
Molecular Formula HgI2
Molecular Weight (g/mol) 454.39
InChI Key YFDLHELOZYVNJE-UHFFFAOYSA-L
Synonym mercury diiodide, mercuric iodide, mercury ii iodide, red mercuric iodide, mercuric iodide, red, mercury biniodide, mercury iodide, mercurius bijodatus, hydrargyrum diodatum, hydrargyrum bijodatum
PubChem CID 24485
ChEBI CHEBI:49659
IUPAC Name diiodomercury
SMILES I[Hg]I

Specifications

Boiling Point 354.0°C
Melting Point 259.0°C
Quantity 500 g
Linear Formula HgI2
Fieser 02,50; 03,7
Merck Index 15,5946
Solubility Information (In potassium iodide soln.) Passes test
Formula Weight 454.39
Grade ACS Reagent
Packaging Glass Bottle
Chemical Name or Material Mercury(II) iodide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463319 Certificate TypeCertificate of Analysis Date30/05/2024 Catalog Number193501000,193500250,193505000
Lot NumberA0459286 Certificate TypeCertificate of Analysis Date16/01/2024 Catalog Number193501000,193500250,193505000
Lot NumberA0392985 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number193501000,193500250,193505000
Lot NumberA0409798 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number193501000,193500250,193505000
Lot NumberA0417351 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number193501000,193500250,193505000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Mercury(II) iodide, ACS reagent, Red, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.