missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mercury(Ii) Iodide, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Quantity | 500 g |
| Linear Formula | HgI2 |
| UN Number | 1638 |
| Solubility Information | Soluble in Potassium Iodide solution |
| Formula Weight | 454.4g/mol |
| Percent Purity | ≥99% |
| Grade | ACS Reagent |
| Chemical Name or Material | Mercury(Ii) Iodide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction