missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) iodide, red, 99+%, Thermo Scientific Chemicals

£24.72 - £1903.47

Chemical Identifiers

CAS 7774-29-0
Molecular Formula HgI2
Molecular Weight (g/mol) 454.39
MDL Number MFCD00011044
InChI Key YFDLHELOZYVNJE-UHFFFAOYSA-L
Synonym mercury diiodide, mercuric iodide, mercury ii iodide, red mercuric iodide, mercuric iodide, red, mercury biniodide, mercury iodide, mercurius bijodatus, hydrargyrum diodatum, hydrargyrum bijodatum
PubChem CID 24485
ChEBI CHEBI:49659
IUPAC Name diiodomercury
SMILES I[Hg]I
View More Specs

Alert:

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.


Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10175070
View Documents
Thermo Scientific Acros
205740050
5 g Glass bottle
£24.72
5g
Please to purchase this item. Need a web account? Register with us today!
10236550
View Documents
Thermo Scientific Acros
205741000
100 g Glass bottle
£82.51
100g
Please to purchase this item. Need a web account? Register with us today!
10328010
View Documents
Thermo Scientific Acros
205745000
500 g Glass bottle
£365.82
500g
Please to purchase this item. Need a web account? Register with us today!
10608514
View Documents
Thermo Scientific Acros
205740025
2.5 kg Glass bottle
£1903.47
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7774-29-0
454.39
YFDLHELOZYVNJE-UHFFFAOYSA-L
24485
diiodomercury
HgI2
MFCD00011044
mercury diiodide, mercuric iodide, mercury ii iodide, red mercuric iodide, mercuric iodide, red, mercury biniodide, mercury iodide, mercurius bijodatus, hydrargyrum diodatum, hydrargyrum bijodatum
CHEBI:49659
I[Hg]I

Specifications

354.0°C
7774-29-0
Crystalline Powder
HgI2
MFCD00011044
15, 5946
(in Potassium iodide soln.) Passes test
I[Hg]I
454.39
CHEBI:49659
≥99%
Mercury(II) iodide, red, p.a.
259.0°C
Red
99% min. (Iodometry)
HgI2
02,50; 03,70
mercury diiodide, mercuric iodide, mercury ii iodide, red mercuric iodide, mercuric iodide, red, mercury biniodide, mercury iodide, mercurius bijodatus, hydrargyrum diodatum, hydrargyrum bijodatum
YFDLHELOZYVNJE-UHFFFAOYSA-L
diiodomercury
24485
454.39
6.36
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Very toxic to aquatic life with long lasting effects.
May cause damage to organs through prolonged or repeated exposure.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 231-873-8

missing translation for 'rtecsNumber' : OW5250000

missing translation for 'tsca' : TSCA

Product Identifier
  • Mercury(II) iodide

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 1
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1

Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P273-Avoid release to the environment.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.