Learn More
Mercury(II) nitrate monohydrate, 98+%, Thermo Scientific Chemicals
£44.32 - £547.73
Chemical Identifiers
CAS | 7783-34-8 |
---|---|
Molecular Formula | H2HgN2O7 |
Molecular Weight (g/mol) | 342.62 |
MDL Number | MFCD00149736 |
InChI Key | KVICROHOONHSRH-UHFFFAOYSA-N |
Synonym | mercuric nitrate monohydrate, unii-yj09edi5uy, mercury ii nitrate hydrate, yj09edi5uy, mercury 2+ ion hydrate dinitrate, nitric acid, mercury 2+ salt, monohydrate, mercury nitrate monohydrate, 324.60 anhydrous, mercury 2+ dinitrate hydrate, hgn2o6.xh2o x=1-2 |
PubChem CID | 3084029 |
IUPAC Name | mercury(2+) hydrate dinitrate |
SMILES | O.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Alert:
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10145080
|
Thermo Scientific Acros
213140500 |
50 g | Plastic bottle |
£44.32
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10115130
|
Thermo Scientific Acros
213142500 |
250 g | Plastic bottle |
£161.81
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10195733
|
Thermo Scientific Acros
213140010 |
1 kg | Plastic bottle |
£547.73
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7783-34-8 | |
342.62 | |
KVICROHOONHSRH-UHFFFAOYSA-N | |
3084029 | |
O.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
H2HgN2O7 | |
MFCD00149736 | |
mercuric nitrate monohydrate, unii-yj09edi5uy, mercury ii nitrate hydrate, yj09edi5uy, mercury 2+ ion hydrate dinitrate, nitric acid, mercury 2+ salt, monohydrate, mercury nitrate monohydrate, 324.60 anhydrous, mercury 2+ dinitrate hydrate, hgn2o6.xh2o x=1-2 | |
mercury(2+) hydrate dinitrate |
Specifications
77.0°C to 79.0°C | |
White | |
98% min. (Complexometry) | |
Hg(NO3)2·H2O | |
03,197; 10,254; 11,317 | |
mercuric nitrate monohydrate, unii-yj09edi5uy, mercury ii nitrate hydrate, yj09edi5uy, mercury 2+ ion hydrate dinitrate, nitric acid, mercury 2+ salt, monohydrate, mercury nitrate monohydrate, 324.60 anhydrous, mercury 2+ dinitrate hydrate, hgn2o6.xh2o x=1-2 | |
KVICROHOONHSRH-UHFFFAOYSA-N | |
mercury(2+) hydrate dinitrate | |
3084029 | |
≥98% | |
4.3000g/mL |
7783-34-8 | |
Adhering Crystalline Powder | |
H2HgN2O7 | |
MFCD00149736 | |
14, 5880 | |
Solubility in water: slightly soluble | |
O.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
342.62 | |
342.6 | |
4.3 | |
Mercury(II) nitrate monohydrate, p.a. |
Safety and Handling
GHS H Statement
May cause damage to organs through prolonged or repeated exposure.
Fatal if swallowed.
Very toxic to aquatic life with long lasting effects.
Fatal in contact with skin.
Fatal if inhaled.
GHS P Statement
Rinse mouth.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Re
GHS Signal Word: Danger
- Mercury(II) nitrate monohydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 1
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H310-Fatal in contact with skin.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H330-Fatal if inhaled.
- H373-May cause damage to organs.
- H300-Fatal if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P330-Rinse mouth.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.