Learn More
Mercury(II) oxide, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Description
Mercury(II) oxide is used in marine and porcelain paints, dry batteries, chemical reagents, and topical antiseptics. It acts as a significant part in the production of mercury. It is utilitarian in the deduction of a mercury based superconductor. It is likewise employed, as a photo-oxidant for the photochemical oxidation of primary aromatic amines to the corresponding azoaromatic compounds. It is also an essential material for the preparation of cathodes in mercury batteries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Orange |
Physical Form | Powder |
Quantity | 5 g |
Assay Percent Range | 99.998% (metals basis) |
Formula Weight | 216.59 |
Odor | Odorless |
Appearance | Dark orange powder |
Chemical Name or Material | Mercury(II) oxide |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
X19I045 | Certificate of Analysis | 28/10/2022 | 010808.14,010808.36,010808.22,010810.06 |
P11A061 | Certificate of Analysis | 12/11/2021 | 010808.14,010808.36,010808.22,010810.06 |
D15Z031 | Certificate of Analysis | 12/11/2021 | 010808.14,010808.36,010808.22,010810.06 |
K18X018 | Certificate of Analysis | 12/11/2021 | 010808.14,010808.36,010808.22,010810.06 |
23157 | Certificate of Analysis | 12/11/2021 | 010808.14,010808.36,010808.22,010810.06 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.