missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) oxide, red, 99%, Thermo Scientific Chemicals

£31.36 - £289.44

Chemical Identifiers

CAS 21908-53-2
Molecular Formula HgO
Molecular Weight (g/mol) 216.59
MDL Number MFCD00011045
InChI Key UKWHYYKOEPRTIC-UHFFFAOYSA-N
Synonym mercuric oxide, mercury ii oxide, mercury oxide, mercuric oxide, red, red mercuric oxide, mercuric oxide, yellow, santar, red precipitate, mercury monoxide
IUPAC Name oxomercury
SMILES O=[Hg]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11423217
View Documents
Thermo Scientific Alfa Aesar
A16157.18
50 g
£31.36
50g
Please to purchase this item. Need a web account? Register with us today!
11433217
View Documents
Thermo Scientific Alfa Aesar
A16157.30
250 g
£100.30
250g
Please to purchase this item. Need a web account? Register with us today!
11413217
View Documents
Thermo Scientific Alfa Aesar
A16157.0B
1000 g
£289.44
1000g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Mercury(II) oxide is used in marine and porcelain paints, dry batteries, chemical reagents, and topical antiseptics. It acts as a significant part in the production of mercury. It is utilitarian in the deduction of a mercury based superconductor. It is likewise employed, as a photo-oxidant for the photochemical oxidation of primary aromatic amines to the corresponding azoaromatic compounds. It is also an essential material for the preparation of cathodes in mercury batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mercury(II) oxide is used in marine and porcelain paints, dry batteries, chemical reagents, and topical antiseptics. It acts as a significant part in the production of mercury. It is utilitarian in the deduction of a mercury based superconductor. It is likewise employed, as a photo-oxidant for the photochemical oxidation of primary aromatic amines to the corresponding azoaromatic compounds. It is also an essential material for the preparation of cathodes in mercury batteries.

Solubility
Partially soluble in water. insoluble in alcohol, ether, acetone and ammonia.

Notes
Light sensitive. Incompatible with hypophosphoric acid, fuels, strong oxidizing agents, strong reducing agents, combustible materials, organic materials and phenols.
Specifications

Chemical Identifiers

21908-53-2
216.59
UKWHYYKOEPRTIC-UHFFFAOYSA-N
oxomercury
HgO
MFCD00011045
mercuric oxide, mercury ii oxide, mercury oxide, mercuric oxide, red, red mercuric oxide, mercuric oxide, yellow, santar, red precipitate, mercury monoxide
O=[Hg]

Specifications

∼500°C (decomposition)
HgO
UN1641
mercuric oxide, mercury ii oxide, mercury oxide, mercuric oxide, red, red mercuric oxide, mercuric oxide, yellow, santar, red precipitate, mercury monoxide
UKWHYYKOEPRTIC-UHFFFAOYSA-N
oxomercury
216.59
Odorless
Mercury(II) oxide, red
21908-53-2
MFCD00011045
14,5882
Partially soluble in water. insoluble in alcohol,ether,acetone and ammonia.
O=[Hg]
216.59
99%
11.14
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.

P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c

H300+H310+H330-H373

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MERCURY OXIDE

missing translation for 'einecsNumber' : 244-654-7

missing translation for 'rtecsNumber' : OW8750000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only