missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) oxide, red, ACS, 99.0% min

Product Code. p-4781995
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
This item is not returnable. View return policy

Product Code. 11371619

Brand: Thermo Scientific Alfa Aesar 012277.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 21908-53-2 | HgO | 216.59 g/mol

It is used in marine and porcelain paints, dry batteries, chemical reagents, and topical antiseptics. It acts as a significant part in the production of mercury. It is utilitarian in the deduction of a mercury based superconductor. It is likewise employed, as a photo-oxidant for the photochemical oxidation of primary aromatic amines to the corresponding azoaromatic compounds. It is also an essential material for the preparation of cathodes in mercury batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21908-53-2
Molecular Formula HgO
Molecular Weight (g/mol) 216.59
MDL Number MFCD00011045
InChI Key UKWHYYKOEPRTIC-UHFFFAOYSA-N
Synonym mercuric oxide, mercury ii oxide, mercury oxide, mercuric oxide, red, red mercuric oxide, mercuric oxide, yellow, santar, red precipitate, mercury monoxide
IUPAC Name oxomercury
SMILES O=[Hg]

Specifications

Color Orange-Red
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.0% min.
Formula Weight 216.59
Grade ACS
Odor Odorless
Appearance Dark orange powder
Chemical Name or Material Mercury(II) oxide, red

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.