missing translation for 'onlineSavingsMsg'
Learn More
Mercury(II) potassium iodide, Thermo Scientific Chemicals Product Code.: 11373718

Mercury(II) potassium iodide, Thermo Scientific Chemicals

Product Code. 11373718
250 g
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy

Product Code. 11373718

Brand: Thermo Scientific Alfa Aesar 039307.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mercury(II) potassium iodide is used as a pharmaceutical intermediate. It is also used as reagents, chemical research and organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-33-7
Molecular Formula H4HgI4K2
Molecular Weight (g/mol) 790.44
MDL Number MFCD00049656
InChI Key ZCPVFAXYZYZMBC-UHFFFAOYSA-N
Synonym nessler's reagent, nessler reagent, mayer's reagent, channing's solution, caswell no. 695, potassium iodomercurate, mercury ii potassium iodide, mercury potassium iodide, potassium mercuric iodide, mercuric potassium iodide
PubChem CID 24542
ChEBI CHEBI:51568
IUPAC Name dipotassium;tetraiodomercury(2-)
SMILES [K+].[K+].I.I.I.I.[Hg--]

Specifications

Color Yellow
Physical Form Crystalline Aggregates
Quantity 250 g
Linear Formula K2HgI4
UN Number UN2025
Merck Index 14,7688
Solubility Information Soluble in water.
Formula Weight 786.4
Sensitivity Hygroscopic
Density 4.3 g/mL
Chemical Name or Material Mercury(II) potassium iodide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ01A035 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number039307.18,039307.30
Lot NumberB08Z022 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number039307.18,039307.30
Lot NumberK29S007 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number039307.18,039307.30
Lot NumberB10N09 Certificate TypeCertificate of Analysis Date29/11/2021 Catalog Number039307.18,039307.30
Lot NumberA03T015 Certificate TypeCertificate of Analysis Date29/11/2021 Catalog Number039307.18,039307.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Mercury(II) potassium iodide, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.