missing translation for 'onlineSavingsMsg'
Learn More
Mercury(II) sulfate, ACS, 98.0% min, Thermo Scientific Chemicals Product Code.: 11363358

Mercury(II) sulfate, ACS, 98.0% min, Thermo Scientific Chemicals

Product Code. 11363358
500 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11363358

Brand: Thermo Scientific Alfa Aesar 036286.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For Use As a Primary Battery Electrolyte, Catalyst in Conversion of Acetylene To Acetaldehyde

Mercury(II) sulfate is used for gold and silver extraction, and to make other mercury compounds. Used as catalyst in the conversion of acetylene to acetaldehyde and used as a electrolyte in primary battery. It is commonly used as a catalyst in oxymercuration-demercuration, a type of electrophilic addition reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-35-9
Molecular Formula HgO4S
Molecular Weight (g/mol) 296.65
MDL Number MFCD00011047
InChI Key DOBUSJIVSSJEDA-UHFFFAOYSA-L
Synonym mercury ii sulfate, mercuric sulfate, mercury sulphate, mercury bisulfate, mercury disulfate, mercury persulfate, mercuric sulphate, mercury sulfate hgso4, unii-j4l3ppg58i, mercuric bisulphateShow MoreShow Less
PubChem CID 24544
IUPAC Name mercury(2+);sulfate
SMILES [Hg++].[O-]S([O-])(=O)=O

Specifications

Melting Point (decomposition)
Color White to Yellow
Physical Form Powder
Quantity 500 g
Assay Percent Range 98% min.
Solubility Information Soluble in hot sulfuric acid and sodium chloride solution. Insoluble in alcohol,acetone and ammonia.
Formula Weight 296.65
Grade ACS
Sensitivity Hygroscopic
Appearance White to pale yellow powder
Density 6.47
Chemical Name or Material Mercury(II) sulfate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX16F008 Certificate TypeCertificate of Analysis Date15/01/2025 Catalog Number036286.22,036286.14,036286.36
Lot NumberN05K044 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number036286.22,036286.14,036286.36
Lot NumberM12K012 Certificate TypeCertificate of Analysis Date19/01/2024 Catalog Number036286.22,036286.14,036286.36
Lot NumberM11K035 Certificate TypeCertificate of Analysis Date18/01/2024 Catalog Number036286.22,036286.14,036286.36
Lot NumberQ17J023 Certificate TypeCertificate of Analysis Date26/04/2023 Catalog Number036286.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Mercury(II) sulfate, ACS, 98.0% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.