missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) sulfate, ACS, 98.0% min

Product Code. p-4781996
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11343358

Brand: Thermo Scientific Alfa Aesar 036286.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For Use As a Primary Battery Electrolyte, Catalyst in Conversion of Acetylene To Acetaldehyde | CAS: 7783-35-9 | HgO4S | 296.65 g/mol

Mercury(II) sulfate is used for gold and silver extraction, and to make other mercury compounds. Used as catalyst in the conversion of acetylene to acetaldehyde and used as a electrolyte in primary battery. It is commonly used as a catalyst in oxymercuration-demercuration, a type of electrophilic addition reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-35-9
Molecular Formula HgO4S
Molecular Weight (g/mol) 296.65
MDL Number MFCD00011047
InChI Key DOBUSJIVSSJEDA-UHFFFAOYSA-L
Synonym mercury ii sulfate, mercuric sulfate, mercury sulphate, mercury bisulfate, mercury disulfate, mercury persulfate, mercuric sulphate, mercury sulfate hgso4, unii-j4l3ppg58i, mercuric bisulphate
PubChem CID 24544
IUPAC Name mercury(2+);sulfate
SMILES [Hg++].[O-]S([O-])(=O)=O

Specifications

Melting Point (decomposition)
Color White to Yellow
Physical Form Powder
Quantity 25 g
Assay Percent Range 98% min.
Solubility Information Soluble in hot sulfuric acid and sodium chloride solution. Insoluble in alcohol,acetone and ammonia.
Formula Weight 296.65
Grade ACS
Sensitivity Hygroscopic
Appearance White to pale yellow powder
Density 6.470 g/cm3 at 20°C
Chemical Name or Material Mercury(II) sulfate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.