missing translation for 'onlineSavingsMsg'
Learn More
Mercury(II) sulfate, ACS reagent, Thermo Scientific Chemicals Product Code.: 10311451

Mercury(II) sulfate, ACS reagent, Thermo Scientific Chemicals

Product Code. 10311451
100 g, Plastic bottle
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Product Code. 10311451

Brand: Thermo Scientific Acros 413651000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-35-9
Molecular Formula HgO4S
Molecular Weight (g/mol) 296.65
MDL Number MFCD00011047
InChI Key DOBUSJIVSSJEDA-UHFFFAOYSA-L
Synonym mercury ii sulfate, mercuric sulfate, mercury sulphate, mercury bisulfate, mercury disulfate, mercury persulfate, mercuric sulphate, mercury sulfate hgso4, unii-j4l3ppg58i, mercuric bisulphateShow MoreShow Less
PubChem CID 24544
IUPAC Name mercury(2+);sulfate
SMILES [Hg++].[O-]S([O-])(=O)=O

Specifications

Color White
Physical Form Crystalline Powder
Quantity 100 g
Linear Formula HgSO4
Fieser 01,658; 15,200
Merck Index 15, 5955
Solubility Information Solubility in water: reacts. Other solubilities: soluble in hcl, hot dil. h2so4 and sol. of nacl
Formula Weight 296.64
Percent Purity ≥98.0%
Grade ACS Reagent
Packaging Plastic bottle
Specific Gravity 6.47
Density 6.4700g/mL
Chemical Name or Material Mercury(II) sulfate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0402008 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number413651000,413655000,413650025
Lot NumberA0407872 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number413651000,413655000,413650025
Lot NumberA0414002 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number413651000,413655000,413650025
Lot NumberA0445340 Certificate TypeCertificate of Analysis Date30/11/2022 Catalog Number413651000,413655000,413650025
Lot NumberA0441096 Certificate TypeCertificate of Analysis Date18/04/2022 Catalog Number413651000,413655000,413650025
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Mercury(II) sulfate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H310-Fatal in contact with skin.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Mercury(II) sulfate, ACS reagent, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.