missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) Sulfate, Certified AR for Analysis, Fisher Chemical

Product Code. 11402854
250 g, Plastic powder jar
Click to view available options
Quantity:
250 g
Packaging:
Plastic powder jar
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Specifications

CAS 7783-35-9
Color White
pH 1
Physical Form Powder/Solid
Quantity 250 g
Molecular Formula HgO4S
MDL Number MFCD00011047
Synonym mercury ii sulfate, mercuric sulfate, mercury sulphate, mercury bisulfate, mercury disulfate, mercury persulfate, mercuric sulphate, mercury sulfate hgso4, unii-j4l3ppg58i, mercuric bisulphate
InChI Key DOBUSJIVSSJEDA-UHFFFAOYSA-L
SMILES [Hg++].[O-]S([O-])(=O)=O
IUPAC Name mercury(2+);sulfate
Molecular Weight (g/mol) 296.65
PubChem CID 24544
Formula Weight 296.64g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Mercury(II) sulfate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H310-Fatal in contact with skin.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P350-IF ON SKIN: Gently wash with plenty of soap and water.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/A§.
  • P330-Rinse mouth.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Mercury(II) Sulfate, Certified AR for Analysis, Fisher Chemical >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.