missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) Sulfate, Extra Pure, SLR, Fisher Chemical™

£201.00 - £478.00

Chemical Identifiers

CAS 7783-35-9
Molecular Formula HgO4S
Molecular Weight (g/mol) 296.65
MDL Number MFCD00011047
InChI Key DOBUSJIVSSJEDA-UHFFFAOYSA-L
Synonym mercury ii sulfate, mercuric sulfate, mercury sulphate, mercury bisulfate, mercury disulfate, mercury persulfate, mercuric sulphate, mercury sulfate hgso4, unii-j4l3ppg58i, mercuric bisulphate
PubChem CID 24544
IUPAC Name mercury(2+);sulfate
SMILES [Hg++].[O-]S([O-])(=O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10687412
View Documents
Fisher Chemical
M/3200/48
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10060240
View Documents
Fisher Chemical
M/3200/50
250 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

7783-35-9
296.65
DOBUSJIVSSJEDA-UHFFFAOYSA-L
24544
[Hg++].[O-]S([O-])(=O)=O
HgO4S
MFCD00011047
mercury ii sulfate, mercuric sulfate, mercury sulphate, mercury bisulfate, mercury disulfate, mercury persulfate, mercuric sulphate, mercury sulfate hgso4, unii-j4l3ppg58i, mercuric bisulphate
mercury(2+);sulfate

Specifications

7783-35-9
1
HgO4S
mercury ii sulfate, mercuric sulfate, mercury sulphate, mercury bisulfate, mercury disulfate, mercury persulfate, mercuric sulphate, mercury sulfate hgso4, unii-j4l3ppg58i, mercuric bisulphate
[Hg++].[O-]S([O-])(=O)=O
296.65
296.64g/mol
White
Powder/Solid
MFCD00011047
DOBUSJIVSSJEDA-UHFFFAOYSA-L
mercury(2+);sulfate
24544
Plastic powder jar
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Mercury(II) sulfate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H310-Fatal in contact with skin.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H300-Fatal if swallowed.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P350-IF ON SKIN: Gently wash with plenty of soap and water.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/A§.
  • P330-Rinse mouth.

SDS
Documents

Documents

Product Certifications