missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mercury(II) sulfate, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
10 g
50 g
100 g
250 g
500 g
3 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
Specifications
Quantity | 100 g |
Linear Formula | HgSO4 |
UN Number | UN1645 |
Formula Weight | 296.65g/mol |
Percent Purity | ≥99% (precipitation titration) |
Chemical Name or Material | Mercury(II) sulfate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Mercury(II) sulfate, Honeywell Fluka™ >
Spot an opportunity for improvement?Share a Content Correction