missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) sulfate, Honeywell Fluka™

Product Code. 15657280
Click to view available options
Quantity:
10 g
50 g
100 g
250 g
500 g
3 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Puriss. p.a., ACS Reagent,≥99.0% (precipitation titration)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-35-9
Molecular Formula HgO4S
Molecular Weight (g/mol) 296.65
MDL Number MFCD00011047
InChI Key DOBUSJIVSSJEDA-UHFFFAOYSA-L
Synonym mercury ii sulfate, mercuric sulfate, mercury sulphate, mercury bisulfate, mercury disulfate, mercury persulfate, mercuric sulphate, mercury sulfate hgso4, unii-j4l3ppg58i, mercuric bisulphate
PubChem CID 24544
IUPAC Name mercury(2+);sulfate
SMILES [Hg++].[O-]S([O-])(=O)=O

Specifications

Quantity 10 g
Linear Formula HgSO4
UN Number UN1645
Formula Weight 296.65g/mol
Percent Purity ≥99% (precipitation titration)
Chemical Name or Material Mercury(II) sulfate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.