missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mercury(II) sulfate, Honeywell Fluka™
Click to view available options
Quantity:
10 g
50 g
100 g
250 g
500 g
3 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 3 kg |
Linear Formula | HgSO4 |
UN Number | UN1645 |
Formula Weight | 296.65g/mol |
Percent Purity | ≥99% (precipitation titration) |
Chemical Name or Material | Mercury(II) sulfate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'