missing translation for 'onlineSavingsMsg'
Learn More
Mercury(II) telluride, 99.999% (metals basis), Thermo Scientific Chemicals Product Code.: 11374177

Mercury(II) telluride, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11374177
10 g
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy

Product Code. 11374177

Brand: Thermo Scientific Alfa Aesar 036666.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mercury cadmium telluride (MCT)is the mostim- portantmaterial currently in use for the manufacture of high performance infrared (IR) detectors. Mercury telluride and mercury telluride-cadmium telluride are semiconductors which have been investigated as intrinsic infrared detecting materials. Complete coverage of the low-loss fiber windows by mercury telluride quantum dot are used in colloidal nanocrystals for telecommunications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12068-90-5
Molecular Formula HgTe
Molecular Weight (g/mol) 328.19
MDL Number MFCD00016133
InChI Key VCEXCCILEWFFBG-UHFFFAOYSA-N
Synonym mercury telluride, mercury ii telluride, mercuric telluride, quecksilbertellurid, mercurous telluride, mercury ii telluride trace metals basis 5g
PubChem CID 82914
IUPAC Name tellanylidenemercury
SMILES [Te]=[Hg]

Specifications

Melting Point 673°C
Color Black to Gray
Physical Form Fused Lump
Quantity 10 g
UN Number UN2025
Solubility Information Insoluble in water.
Formula Weight 328.19
Percent Purity 99.999%
Odor Odorless
Density 8.63 g/mL
Assay (metals basis)
Chemical Name or Material Mercury(II) telluride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberB18U025 Certificate TypeCertificate of Analysis Date28/12/2021 Catalog Number036666.04,036666.09
Lot NumberR12H011 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number036666.04,036666.09
Lot NumberN25G027 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number036666.04,036666.09
Lot NumberH13S019 Certificate TypeCertificate of Analysis Date29/11/2021 Catalog Number036666.04,036666.09
Lot NumberB10U038 Certificate TypeCertificate of Analysis Date29/11/2021 Catalog Number036666.04,036666.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Mercury(II) telluride, 99.999% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.