missing translation for 'onlineSavingsMsg'
Learn More
Mercury(II) thiocyanate, Thermo Scientific Chemicals Product Code.: 11394277

Mercury(II) thiocyanate, Thermo Scientific Chemicals

Product Code. 11394277
50 g
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Product Code. 11394277

Brand: Thermo Scientific Alfa Aesar 037106.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Mercury(II) thiocyanate is used as a precursor to potassium tris(thiocyanato)mercurate(II) and cesium tris(thiocyanato)mercurate(II). It is also used in the determination of chloride ions in water by UV-visible spectroscopy. Further, it acts as a catalyst for the addition of thiocyanic acid to alkynes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 592-85-8
Molecular Formula C4HgK2N4S4
Molecular Weight (g/mol) 511.10
MDL Number MFCD00011039
InChI Key HWXOCCNUUYRBKP-UHFFFAOYSA-J
IUPAC Name mercury(2+) dipotassium tetrakis(cyanosulfanide)
SMILES [K+].[K+].[Hg++].[S-]C#N.[S-]C#N.[S-]C#N.[S-]C#N

Specifications

Melting Point ∽165°C (decomposition)
Color White to Yellow
Physical Form Powder
Quantity 50 g
Beilstein 3561344
Solubility Information Soluble in dilute hydrochloric acid,potassium cyanide and ammonia. Slightly soluble in alcohol and ether. Insoluble in water.
Formula Weight 316.75
Odor Odorless
Sensitivity Hygroscopic
Appearance White to pale yellow powder
Density 3.718 g/cm3 at 20°C
Chemical Name or Material Mercury(II) thiocyanate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP03L002 Certificate TypeCertificate of Analysis Date05/03/2025 Catalog Number037106.18,037106.30
Lot NumberX11K028 Certificate TypeCertificate of Analysis Date05/11/2024 Catalog Number037106.18,037106.30
Lot NumberQ03K049 Certificate TypeCertificate of Analysis Date17/04/2024 Catalog Number037106.18,037106.30
Lot NumberN07K039 Certificate TypeCertificate of Analysis Date13/02/2024 Catalog Number037106.18,037106.30
Lot NumberM23J020 Certificate TypeCertificate of Analysis Date27/01/2023 Catalog Number037106.18,037106.30
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Mercury(II) thiocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
Supplemental information
  • MIXTURE LIST-Contains : Mercuric thiocyanate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Mercury(II) thiocyanate, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.