missing translation for 'onlineSavingsMsg'
Learn More

Mercury standard solution, for AAS, solution, 1 mg/ml Hg in 10% HNO3, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 196121000

15.17 GBP valid until 2024-12-20
Use promo code "21695" to get your promotional price.



 View more versions of this product

Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 10710901

  • £21.11 / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Alert:

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For atomic adsorption spectroscopy
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Mercury standard solution
1 mg/ml Hg in 10% HNO3
Plastic bottle
Black to Gray
MFCD00149736
O.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
342.62
342.62
1.0200g/mL
101.0°C
Soluble in water
990 to 1010μg/mL of Hg(2+) in 2 to 5% HNO3
7783-34-8, 7697-37-2, 7732-18-5
100 mL
H2HgN2O7
KVICROHOONHSRH-UHFFFAOYSA-N
mercury(2+);dinitrate;hydrate
3084029
AAS
-3.0°C
1.02
Powder, Granules or Chunks
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Mercury atomic absorption standard solution, 1 mg/ml Hg in 10% HNO3

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H373-May cause damage to organs.
  • H290-May be corrosive to metals.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only