Learn More
Thermo Scientific Chemicals MES, 99+%, <1% water
£94.60 - £1033.00
Chemical Identifiers
CAS | 4432-31-9 |
---|---|
Molecular Formula | C6H13NO4S |
Molecular Weight (g/mol) | 195.233 |
MDL Number | MFCD00006181 |
InChI Key | SXGZJKUKBWWHRA-UHFFFAOYSA-N |
Synonym | 2-(N-Morpholino)ethanesulfonic acid |
PubChem CID | 78165 |
ChEBI | CHEBI:39005 |
IUPAC Name | 2-morpholin-4-ylethanesulfonic acid |
SMILES | C1COCCN1CCS(=O)(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10254904
|
Thermo Scientific Chemicals
274140500 |
50 g | Glass bottle |
£94.60
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10413004
|
Thermo Scientific Chemicals
274142500 |
250 g | Glass bottle |
£323.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10726085
|
Thermo Scientific Chemicals
274140010 |
1 kg | Plastic bottle |
£1033.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferChemical Identifiers
4432-31-9 | |
195.233 | |
SXGZJKUKBWWHRA-UHFFFAOYSA-N | |
78165 | |
2-morpholin-4-ylethanesulfonic acid |
C6H13NO4S | |
MFCD00006181 | |
2-(N-Morpholino)ethanesulfonic acid | |
CHEBI:39005 | |
C1COCCN1CCS(=O)(=O)O |
Specifications
4432-31-9 | |
White | |
99+% | |
C6H13NO4S | |
15, 5972 | |
Solubility in water: 10g/100mL. | |
C1COCCN1CCS(=O)(=O)O | |
195.233 | |
CHEBI:39005 | |
≥99% |
>300.0°C | |
Powder | |
Authentic | |
MFCD00006181 | |
2-(N-Morpholino)ethanesulfonic acid | |
SXGZJKUKBWWHRA-UHFFFAOYSA-N | |
2-morpholin-4-ylethanesulfonic acid | |
78165 | |
195.24 | |
MES |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash face, hands and any exposed skin thoroughly after handling.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Use only outdoors or in a well-ventilated area.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RUO – Research Use Only