missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals MES monohydrate, 98% Product Code.: 11433137

Thermo Scientific Chemicals MES monohydrate, 98%

Product Code. 11433137
500 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11433137

Brand: Thermo Scientific Chemicals A16104.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

MES monohydrate is a biological buffer often referred to as Good’s buffer. It is useful for cell culture media and protein based buffer formulations to maintain a stable environment in solution. It is also used in the purification bioprocesses of antibodies, peptides, proteins, blood components and growth factors. It is a useful resolving agent for very small proteins on bis-tris gels. Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
MES monohydrate is a biological buffer often referred to as Good’s buffer. It is useful for cell culture media and protein based buffer formulations to maintain a stable environment in solution. It is also used in the purification bioprocesses of antibodies, peptides, proteins, blood components and growth factors. It is a useful resolving agent for very small proteins on bis-tris gels. Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145224-94-8
Molecular Formula C6H15NO5S
Molecular Weight (g/mol) 213.248
MDL Number MFCD00149409
InChI Key MIIIXQJBDGSIKL-UHFFFAOYSA-N
PubChem CID 16218417
IUPAC Name 2-morpholin-4-ylethanesulfonic acid;hydrate
SMILES C1COCCN1CCS(=O)(=O)O.O

Specifications

Melting Point ∽308°C (decomposition)
Quantity 500 g
Assay Percent Range 98%
Beilstein 141319
Merck Index 14,5902
Solubility Information Soluble in water.
Formula Weight 213.26 (195.24 Anhydrous)
Percent Purity 98%
Chemical Name or Material MES Monohydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252676 Certificate TypeCertificate of Analysis Date05/10/2024 Catalog NumberA16104.14,A16104.36,A16104.22
Lot Number10248132 Certificate TypeCertificate of Analysis Date28/09/2023 Catalog NumberA16104.14,A16104.36,A16104.22
Lot Number10240887 Certificate TypeCertificate of Analysis Date25/05/2022 Catalog NumberA16104.14,A16104.36,A16104.22
Lot Number10236807 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16104.14,A16104.36,A16104.22
Lot Number10191845 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16104.14,A16104.36,A16104.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals MES monohydrate, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.