Learn More
Thermo Scientific Chemicals MES monohydrate, 99+%, for biochemistry
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Absorbance | (0.1M in H2O) 1 cm cell, (0.1M in H2O) 1 cm cell at 0nm, 0.02 max. at 280nm, 0.05 max. at 260nm |
Absorbance Conditions | (0.1M aq. soln.) |
Melting Point | >300.0°C |
Color | Orange-Yellow to White |
pH | 2.5 to 4.0 (1%, H2O, 25°C) |
Physical Form | Crystals, Flakes or Powder |
Quantity | 500 g |
Assay Percent Range | 99+% |
Infrared Spectrum | Authentic |
Packaging | Plastic bottle |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0442902 | Certificate of Analysis | 31/01/2025 | 327760025,327761000,327765000 |
A0442604 | Certificate of Analysis | 31/01/2025 | 327760025,327761000,327765000 |
A0465753 | Certificate of Analysis | 26/08/2024 | 327760025,327761000,327765000 |
A0457998 | Certificate of Analysis | 20/11/2023 | 327760025,327761000,327765000 |
A0452534 | Certificate of Analysis | 03/05/2023 | 327760025,327761000,327765000 |
Safety and Handling
- MES monohydrate
Special Offers
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.