missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MES monohydrate, free acid, ≥99%, MP Biomedicals™
A buffer using MES free acid can be prepared by titrating the free acid with sodium hydroxide to the desired pH (pKa ± 1). Alternatively, volumes of equimolar MES free acid and sodium or potassium MES can be mixed to attain the desired pH
£147.00 - £319.00
Chemical Identifiers
CAS | 4432-31-9 |
---|---|
Molecular Formula | C6H13NO4S |
Molecular Weight (g/mol) | 195.233 |
MDL Number | MFCD00149409 |
InChI Key | SXGZJKUKBWWHRA-UHFFFAOYSA-N |
Synonym | 2-[N-morpholino]ethanesulfonic acid |
PubChem CID | 78165 |
ChEBI | CHEBI:39005 |
IUPAC Name | 2-morpholin-4-ylethanesulfonic acid |
SMILES | C1COCCN1CCS(=O)(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11456581
|
MP Biomedicals
0219483580 |
100 g |
£147.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11466581
|
MP Biomedicals
0219483583 |
250 g |
£319.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
4432-31-9 | |
195.233 | |
SXGZJKUKBWWHRA-UHFFFAOYSA-N | |
78165 | |
2-morpholin-4-ylethanesulfonic acid |
C6H13NO4S | |
MFCD00149409 | |
2-[N-morpholino]ethanesulfonic acid | |
CHEBI:39005 | |
C1COCCN1CCS(=O)(=O)O |
Specifications
4432-31-9 | |
Crystalline Powder | |
C6H13NO4S | |
2-[N-morpholino]ethanesulfonic acid | |
C1COCCN1CCS(=O)(=O)O | |
195.233 | |
CHEBI:39005 | |
99.6% |
White | |
≥99% | |
MFCD00149409 | |
SXGZJKUKBWWHRA-UHFFFAOYSA-N | |
2-morpholin-4-ylethanesulfonic acid | |
78165 | |
213.26 | |
MES monohydrate |
Safety and Handling
missing translation for 'einecsNumber' : 224-632-3