missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals MES sodium salt, 99%

Product Code. 10776465 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
missing translation for 'unitSize'
100g
25g
500g
This item is not returnable. View return policy

Product Code. 10776465

missing translation for 'mfr': Thermo Scientific Chemicals 397351000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 71119-23-8 | C6H12NNaO4S | 217.215 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71119-23-8
Molecular Formula C6H12NNaO4S
Molecular Weight (g/mol) 217.215
MDL Number MFCD00065473
InChI Key IRHWMYKYLWNHTL-UHFFFAOYSA-M
PubChem CID 23673676
ChEBI CHEBI:62955
IUPAC Name sodium;2-morpholin-4-ylethanesulfonate
SMILES C1COCCN1CCS(=O)(=O)[O-].[Na+]

Specifications

Absorbance 0.03 max. (10% in water) 1 cm cell vs water at 280nm, 0.05 max. (10% in water) 1 cm cell vs water at 260nm
Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 99%
Infrared Spectrum Authentic
Packaging Plastic bottle
Solubility Information Solubility in water: 0.5g/L.
Formula Weight 217.22
Percent Purity ≥98.5% (after ion exchange)
Chemical Name or Material MES sodium salt
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.