missing translation for 'onlineSavingsMsg'
Learn More
Mesitylene, 97%, pure, Thermo Scientific Chemicals Product Code.: 10408540

Mesitylene, 97%, pure, Thermo Scientific Chemicals

Product Code. 10408540
250 mL, Glass bottle
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10408540

Brand: Thermo Scientific Acros 161322500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-67-8
Molecular Formula C9H12
Molecular Weight (g/mol) 120.19
MDL Number MFCD00008538
InChI Key AUHZEENZYGFFBQ-UHFFFAOYSA-N
Synonym mesitylene, sym-trimethylbenzene, benzene, 1,3,5-trimethyl, fleet-x, trimethylbenzol, 3,5-dimethyltoluene, s-trimethylbenzene, hsdb 92, 2,4,6-trimethylbenzene, 1,3,5-trimethyl-benzene
PubChem CID 7947
ChEBI CHEBI:34833
IUPAC Name 1,3,5-trimethylbenzene
SMILES CC1=CC(=CC(=C1)C)C

Specifications

Melting Point -45.0°C
Color Colorless
Density 0.8675g/mL
Boiling Point 163.0°C to 166.0°C
Flash Point 44°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C6H3(CH3)3
Refractive Index 1.4980 to 1.5000
Quantity 250 mL
Beilstein 05, 406
Merck Index 15, 5977
Specific Gravity 0.8675
Solubility Information Solubility in water: 2.9g/L (20°C). Other solubilities: miscible with alcohol,benzene and ether
Viscosity 0.834 mm2/s (20°C)
Formula Weight 120.19
Percent Purity 97%
Grade Pure
Physical Form Liquid
Chemical Name or Material Mesitylene
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463297 Certificate TypeCertificate of Analysis Date11/10/2024 Catalog Number161320010,161320025,161322500
Lot NumberA0459886 Certificate TypeCertificate of Analysis Date06/02/2024 Catalog Number161320010,161320025,161322500
Lot NumberA0459127 Certificate TypeCertificate of Analysis Date09/01/2024 Catalog Number161320010,161320025,161322500
Lot NumberA0457730 Certificate TypeCertificate of Analysis Date10/11/2023 Catalog Number161320010,161320025,161322500
Lot NumberA0456500 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog Number161320010,161320025,161322500
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Mesitylene
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 1,3,5-Trimethylbenzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Mesitylene, 97%, pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.