Learn More
Mesitylene, 98+%, Thermo Scientific Chemicals
Description
Mesitylene is used to make plastics and dyes. It acts as a solvent, ligand in organometallic chemistry and precursor to 2,4,6-trimethylaniline. It is also used as a developer for photopatternable silicones due to its solvent properties in the electronics industry. It is used as an internal standard in nuclear magnetic resonance (NMR) samples due to the presence of three equivalent protons in it. It is involved in the production of trimesic acid and antioxygen, epoxy firming agent and polyester resin stabilizers. Further, it is used as an additive and component in aviation gasoline blends.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Mesitylene is used to make plastics and dyes. It acts as a solvent, ligand in organometallic chemistry and precursor to 2,4,6-trimethylaniline. It is also used as a developer for photopatternable silicones due to its solvent properties in the electronics industry. It is used as an internal standard in nuclear magnetic resonance (NMR) samples due to the presence of three equivalent protons in it. It is involved in the production of trimesic acid and antioxygen, epoxy firming agent and polyester resin stabilizers. Further, it is used as an additive and component in aviation gasoline blends.
Solubility
Partly miscible with water.
Notes
Incompatible with strong oxidizing agents.

Specifications
Specifications
Melting Point | -45°C |
Density | 0.865 |
Boiling Point | 163°C to 165°C |
Flash Point | 44°C (111°F) |
Odor | Unpleasant |
Refractive Index | 1.499 |
Quantity | 500 mL |
UN Number | UN2325 |
Beilstein | 906806 |
Merck Index | 14,5907 |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10250594 | Certificate of Analysis | 30/08/2024 | A11323.AP,A11323.AE,A11323.0F |
10250590 | Certificate of Analysis | 22/04/2024 | A11323.AP,A11323.AE,A11323.0F |
10245613 | Certificate of Analysis | 09/02/2023 | A11323.AP,A11323.AE,A11323.0F |
10244769 | Certificate of Analysis | 25/11/2022 | A11323.AP,A11323.AE,A11323.0F |
10241203 | Certificate of Analysis | 10/06/2022 | A11323.AP,A11323.AE,A11323.0F |
Safety and Handling




- Mesitylene
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H226-Flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H411-Toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : 1,3,5-Trimethylbenzene
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.