missing translation for 'onlineSavingsMsg'
Learn More
Learn More
meso-1,2-Diphenyl-1,2-ethanediol, 95%, Thermo Scientific Chemicals
£16.72 - £288.00
Chemical Identifiers
CAS | 579-43-1 |
---|---|
Molecular Formula | C14H14O2 |
Molecular Weight (g/mol) | 214.26 |
MDL Number | MFCD00064253 |
InChI Key | IHPDTPWNFBQHEB-UHFFFAOYNA-N |
Synonym | meso-hydrobenzoin, meso-1,2-diphenyl-1,2-ethanediol, unii-co9a49a84i, meso-stilbene glycol, 1r,2s-1,2-diphenylethane-1,2-diol, meso-1,2-diphenylethylene glycol, hydrobenzoin, meso, unii-q61g3433lb component |
PubChem CID | 853018 |
ChEBI | CHEBI:50015 |
IUPAC Name | (1R,2S)-1,2-diphenylethane-1,2-diol |
SMILES | OC(C(O)C1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10678063
|
Thermo Scientific Acros
247680010 |
1 g | Glass bottle | ||||||
10156873
|
Thermo Scientific Acros
247680050 |
5 g | Glass bottle |
£64.50
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10628824
|
Thermo Scientific Acros
247680250 |
25 g | Glass bottle |
£288.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
579-43-1 | |
214.26 | |
IHPDTPWNFBQHEB-UHFFFAOYNA-N | |
853018 | |
(1R,2S)-1,2-diphenylethane-1,2-diol |
C14H14O2 | |
MFCD00064253 | |
meso-hydrobenzoin, meso-1,2-diphenyl-1,2-ethanediol, unii-co9a49a84i, meso-stilbene glycol, 1r,2s-1,2-diphenylethane-1,2-diol, meso-1,2-diphenylethylene glycol, hydrobenzoin, meso, unii-q61g3433lb component | |
CHEBI:50015 | |
OC(C(O)C1=CC=CC=C1)C1=CC=CC=C1 |
Specifications
579-43-1 | |
White | |
94% min. (GC) | |
C6H5CH(OH)CH(OH)C6H5 | |
meso-hydrobenzoin, meso-1,2-diphenyl-1,2-ethanediol, unii-co9a49a84i, meso-stilbene glycol, 1r,2s-1,2-diphenylethane-1,2-diol, meso-1,2-diphenylethylene glycol, hydrobenzoin, meso, unii-q61g3433lb component | |
IHPDTPWNFBQHEB-UHFFFAOYNA-N | |
(1R,2S)-1,2-diphenylethane-1,2-diol | |
853018 | |
214.26 | |
Crystalline Powder |
136.0°C to 137.0°C | |
Authentic | |
C14H14O2 | |
MFCD00064253 | |
Solubility in water: insoluble. | |
OC(C(O)C1=CC=CC=C1)C1=CC=CC=C1 | |
214.26 | |
CHEBI:50015 | |
95% | |
meso-1, 2-Diphenyl-1, 2-ethanediol |
RUO – Research Use Only