missing translation for 'onlineSavingsMsg'
Learn More
Learn More
meso-2,3-Dimercaptosuccinic acid, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
It is applied as a selective masking agent and chelating agent for cadmium in EDTA titration of zinc.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is applied as a selective masking agent and chelating agent for cadmium in EDTA titration of zinc.
Solubility
Soluble in water, and ethanol (25 mg/ml, results in colorless to light yellow, clear).
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is applied as a selective masking agent and chelating agent for cadmium in EDTA titration of zinc.
Solubility
Soluble in water, and ethanol (25 mg/ml, results in colorless to light yellow, clear).
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | ∼196°C (decomposition) |
Quantity | 5 g |
Beilstein | 1725150 |
Sensitivity | Air Sensitive |
Merck Index | 14,8865 |
Solubility Information | Soluble in water,and ethanol (25mg/ml,results in colorless to light yellow,clear). |
Formula Weight | 182.22 |
Percent Purity | 97% |
Chemical Name or Material | meso-2,3-Dimercaptosuccinic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'