Learn More
meso-Erythritol, 99%, Thermo Scientific Chemicals
Description
meso-Erythritol is used to support the carbon source growth requirement of Gluconobacter bacteria. It is also used in the chemical analysis of the reactivity of various radicals and radical anions, in encapsulated phase change materials (PCMs) to study its potential use in microelectronics cooling applications, as a reference compound in purification and analytical procedures developed to test fermentation broths.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
meso-Erythritol is used to support the carbon source growth requirement of Gluconobacter bacteria. It is also used in the chemical analysis of the reactivity of various radicals and radical anions, in encapsulated phase change materials (PCMs) to study its potential use in microelectronics cooling applications, as a reference compound in purification and analytical procedures developed to test fermentation broths.
Solubility
Soluble in water (0.1 g/ml).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store at -20°C.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 119°C to 124°C |
Boiling Point | 329°C to 331°C |
Assay Percent Range | 99% |
Quantity | 25 g |
Beilstein | 1719753 |
Merck Index | 14,3675 |
Solubility Information | Soluble in water (0.1g/ml). |
Formula Weight | 122.12 |
Percent Purity | 99% |
Chemical Name or Material | meso-Erythritol |
RUO – Research Use Only