Learn More
Mestranol, MedChemExpress
MedChemExpress Mestranol is an inactive prodrug and becomes biologically active on conversion to ethinyl estradiol (EE). Mestranol acts as an estrogen receptor agonist. Mestranol combines with a progestin in vivo and can be used for the research of menopausal hormone or menstrual disorders.
Brand:  Medchem Express HY-B0390
Encompass_Preferred
Product Code. 16488276
  • £75.02 / 100mg
Explore more special offers
Description
Description
Mestranol is an inactive prodrug and becomes biologically active on conversion to ethinyl estradiol (EE). Mestranol acts as an estrogen receptor agonist. Mestranol combines with a progestin in vivo and can be used for the research of menopausal hormone or menstrual disorders.
Specifications
Specifications
Mestranol
White
C21H26O2
DMSO : 33.33 mg/mL (107.37 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (2.16 mM; Need ultrasonic)
310.43
99.88%
Solid
72-33-3
Research
100 mg
C[C@@]1([C@]2(O)C#C)[C@](CC2)([H])[C@@](CCC3=C4C=CC(OC)=C3)([H])[C@]4([H])CC1
310.43
Research