missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromophenol in Ethanol 1000μg/mL, Fisher Chemical™

Product Code. 12926153
Click to view available options
Quantity:
1 mL
Packaging:
Ampoule
This item is not returnable. View return policy
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Concentration 1000μg/mL
CAS 615-58-7
Packaging Ampoule
Quantity 1 mL
Type Organic Single-component Standard Solution
Molecular Formula C6H4Br2O
InChI Key FAXWFCTVSHEODL-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1Br)Br)O
IUPAC Name 2,4-dibromophenol
Molecular Weight (g/mol) 251.905
PubChem CID 12005
ChEBI CHEBI:34238
Solvent or Matrix Ethanol
Chemical Name or Material 2,4-Dibromophenol
Show More Show Less
compliance-icons
Product Identifier
  • standard solution 2,4-dibromophenol 1000µg/ml in ethanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contains : ethanol, 2,4-dibromophenol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'