missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluorobiphenyl in DCM 100μg/mL, Fisher Chemical™
Click to view available options
Quantity:
1 mL
Specifications
Specifications
Concentration | 100μg/mL |
CAS | 321-60-8 |
Packaging | Ampoule(s) |
Quantity | 1 mL |
Molecular Formula | C12H9F |
Solvent or Matrix | Dichloromethane |
MDL Number | MFCD00000317 |
InChI Key | KLECYOQFQXJYBC-UHFFFAOYSA-N |
SMILES | FC1=CC=CC=C1C1=CC=CC=C1 |
IUPAC Name | 2-fluoro-1,1'-biphenyl |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction