missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metabolite 4-Chloroaniline in DCM 10μg/mL, Fisher Chemical™
Click to view available options
Quantity:
10 mL
Packaging:
Ampoule(s)
Specifications
Concentration | 10μg/mL |
CAS | 106-47-8 |
Packaging | Ampoule(s) |
Quantity | 10 mL |
Molecular Formula | C6H6ClN |
InChI Key | QSNSCYSYFYORTR-UHFFFAOYSA-N |
SMILES | C1=CC(=CC=C1N)Cl |
IUPAC Name | 4-chloroaniline |
Molecular Weight (g/mol) | 127.571 |
PubChem CID | 7812 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'